n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number ACR445720010
Price (MYR)
575.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
575.00
Add to cart
Specifications
Chemical Name or Materialn-Hexane
CAS110-54-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long las
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or d