Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitromethane, 96%, for HPLC, Thermo Scientific Chemicals
CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number ACR445670010
Price (MYR)
813.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
813.00
Add to cart
Chemical Identifiers
CAS673-22-3
IUPAC Name2-hydroxy-4-methoxybenzaldehyde
Molecular FormulaC8H8O3
InChI KeyWZUODJNEIXSNEU-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale brown or pale pink
FormCrystals or powder or crystalline powder
Melting Point (clear melt)39.0-46.0?C
Assay (GC)≥97.5%