Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals
Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals
Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals
Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals
CAS: 75-15- | CS2 | 76.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR445660010
Price (MYR)
1,042.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,042.00
Add to cart
Chemical Identifiers
CAS6976-37-0
IUPAC Name2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC8H19NO5
InChI KeyOWMVSZAMULFTJU-UHFFFAOYSA-N
SMILESOCCN(CCO)C(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Water=<1.0 % (K.F.)
Iron (Fe)=<5 ppm