Ethylene glycol, technical
Ethylene glycol, technical
Ethylene glycol, technical
Ethylene glycol, technical
Thermo Scientific Chemicals
Ethylene glycol, technical
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Catalog NumberQuantity
ACR4442300101 L
Catalog number ACR444230010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7447-41-8
IUPAC Namelithium(1+) chloride
Molecular FormulaClLi
InChI KeyKWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES[Li+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.005% max.
Particle Size-10 Mesh