Paraffin, liquid, technical
Paraffin, liquid, technical
Paraffin, liquid, technical
Paraffin, liquid, technical
Thermo Scientific Chemicals
Paraffin, liquid, technical
CAS: 8012-95-1 | MFCD00131611
Have Questions?
Catalog NumberQuantityPackaging
ACR44418025025 LPlastic Can
Catalog number ACR444180250
Price (MYR)
2,509.00
Add to cart
Quantity:
25 L
Packaging:
Plastic Can
Request bulk or custom format
Price (MYR)
2,509.00
Add to cart
Chemical Identifiers
CAS1572-98-1
IUPAC Nameethyl 2-cyano-2,2-dimethylacetate
Molecular FormulaC7H11NO2
InChI KeyFYGRPGOHQCPZCV-UHFFFAOYSA-N
SMILESCCOC(=O)C(C)(C)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.4075-1.4125 @ 20?C
FormLiquid
Assay (GC)≥96.0%