2-Butanone, technical
2-Butanone, technical
2-Butanone, technical
2-Butanone, technical
2-Butanone, technical
Thermo Scientific Chemicals
2-Butanone, technical
CAS: 78-93-3 | C4H8O | 72.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4441700505 LPlastic Drum
Catalog number ACR444170050
Price (MYR)
578.00
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
578.00
Add to cart
Chemical Identifiers
CAS5351-23-5
IUPAC Name4-hydroxybenzohydrazide
Molecular FormulaC7H8N2O2
InChI KeyZMZGIVVRBMFZSG-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale grey
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%