Agar, Bacteriological grade
Agar, Bacteriological grade
Thermo Scientific Chemicals
Agar, Bacteriological grade
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number ACR443572500
Price (MYR)
565.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
565.00
Add to cart
Chemical Identifiers
CAS577-85-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or pale cream
Melting Point (clear melt)168.0-174.0°C
Assay (HPLC)≥98.0%
FormPowder
Microbial media
RUO – Research Use Only