Agar, Bacteriological grade
Agar, Bacteriological grade
Thermo Scientific Chemicals
Agar, Bacteriological grade
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number ACR443570050
Price (MYR)
5,606.00
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
5,606.00
Add to cart
Chemical Identifiers
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.2 ppm
Nitrate (NO3)=<0.5 ppm
Arsenic (As)=<0.01 ppm
Iron (Fe)=<0.2 ppm
Ammonium (NH4)=<2 ppm
View more
Microbial media
RUO – Research Use Only