Agar, Bacteriological grade
Agar, Bacteriological grade
Thermo Scientific Chemicals
Agar, Bacteriological grade
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Catalog number ACR443570010
Price (MYR)
1,586.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,586.00
Add to cart
Chemical Identifiers
CAS81-13-0
IUPAC Name(2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
Molecular FormulaC9H19NO4
InChI KeySNPLKNRPJHDVJA-ZETCQYMHSA-N
SMILESCC(C)(CO)[C@@H](O)C(=O)NCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Water Content (Karl Fischer Titration)≤1.0%
Refractive Index1.4950-1.5020 @ 20°C
FormViscous liquid
Identification (FTIR)Conforms
View more
Microbial media
RUO – Research Use Only