2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals
CAS: 19767-45-4 | C2H5NaO3S2 | 164.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number ACR443150100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS7722-76-1
IUPAC Nameammonium dihydrogen phosphate
Molecular FormulaH6NO4P
InChI KeyLFVGISIMTYGQHF-UHFFFAOYSA-N
SMILES[NH4+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
pH4.0 to 5.0 (10 % in water)
UVat 280 nm A: =<0.05
UVat 260 nm A: =<0.05
UV(10 % in water)