Chlorobenzene, 99.5%, Thermo Scientific Chemicals
Chlorobenzene, 99.5%, Thermo Scientific Chemicals
Chlorobenzene, 99.5%, Thermo Scientific Chemicals
Chlorobenzene, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorobenzene, 99.5%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number ACR443001000
Price (MYR)
416.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
416.00
Add to cart
Chemical Identifiers
CAS65039-08-9
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium bromide
Molecular FormulaC6H11BrN2
InChI KeyGWQYPLXGJIXMMV-UHFFFAOYSA-M
SMILES[Br-].CCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Appearance (Color)Yellow
Titration Argentometric>=97.5 %