Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number ACR442981000
Price (MYR)
662.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
662.00
Add to cart
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.4 %
Water=<0.1 % (Coulometric)
Butyl ether=<0.1 %
Substance colored by H2SO4Passes test