Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Catalog number ACR442980010
Price (MYR)
3,828.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
3,828.00
Add to cart
Chemical Identifiers
CAS7791-18-6
IUPAC Namemagnesium(2+) hexahydrate dichloride
Molecular FormulaCl2H12MgO6
InChI KeyDHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Ammonium (NH4)=<50 ppm
Barium (Ba)=<20 ppm
Copper (Cu)=<10 ppm
Potassium (K)=<3000 ppm
Zinc (Zn)=<10 ppm