Acetoacetamide, 97%, Thermo Scientific Chemicals
Acetoacetamide, 97%, Thermo Scientific Chemicals
Acetoacetamide, 97%, Thermo Scientific Chemicals
Acetoacetamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetoacetamide, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR442541000
Price (MYR)
470.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
470.00
Add to cart
Chemical Identifiers
CAS112-39-0
IUPAC Namemethyl hexadecanoate
Molecular FormulaC17H34O2
InChI KeyFLIACVVOZYBSBS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid or low melting solid