Boron nitride, 98%, Thermo Scientific Chemicals
Boron nitride, 98%, Thermo Scientific Chemicals
Boron nitride, 98%, Thermo Scientific Chemicals
Boron nitride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boron nitride, 98%, Thermo Scientific Chemicals
CAS: 10043-11-5 | BN | 24.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number ACR441800500
Price (MYR)
672.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
672.00
Add to cart
Specifications
Chemical Name or MaterialBoron nitride
Melting Point2700.0°C to 3000.0°C
CAS10043-11-5
Quantity50 g
EINECS Number233-136-6