Propidium iodide, 95%
Propidium iodide, 95%
Propidium iodide, 95%
Propidium iodide, 95%
Thermo Scientific Chemicals
Propidium iodide, 95%
CAS: 25535-16-4 | C27H34I2N4 | 668.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 mg
Packaging:
Glass bottle
Catalog number ACR440300010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS327-57-1
IUPAC Name(2S)-2-aminohexanoic acid
Molecular FormulaC6H13NO2
InChI KeyLRQKBLKVPFOOQJ-YFKPBYRVSA-N
SMILESCCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=99.0 %
Other amino acids(TLC) Not detectable
Infrared spectrumConforms
Loss on drying=<1 % (105°C, 4 hrs)