Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals
Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals
Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals
Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals
Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals
CAS: 1793-07-3 | C9H7NO3 | 177.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ACR440150050
Price (MYR)
227.23
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
227.23
Add to cart
Chemical Identifiers
CAS1718-34-9
IUPAC Namesodium (3Z)-3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
Molecular FormulaC13H8N3NaO5
InChI KeyHKSANXZSKDUDBD-SOCRLDLMSA-M
SMILES[Na+].[O-]C(=O)C1=CC(C=CC1=O)=N/NC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color ChangepH 10.1 (yellow)
Color ChangepH 12.1 (orange)