Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
Thermo Scientific Chemicals
Tetrabutylammonium phosphate monobasic, 1M solution in ethanol, AcroSeal™
CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR440061000100 mLAcroSeal™ Glass bottle
Catalog number ACR440061000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-33-7
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Phase transfer catalyst
For Manufacturing and Laboratory Use. Not for beverage use