3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
CAS: 121-00-6 | C11H16O2 | 180.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR439691000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS92136-39-5
IUPAC Nametert-butyl N-(prop-2-yn-1-yl)carbamate
Molecular FormulaC8H13NO2
InChI KeyDSPYCWLYGXGJNJ-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay (GC)>97%
Identification (FTIR)Conforms to structure
Appearance (Color)Yellow to brown