N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%
N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%
N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%
N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%
Thermo Scientific Chemicals
N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%
CAS: 95713-52-3 | C9H9FN4O5 | 272.19 g/mol
Have Questions?
Catalog NumberdimQuantityPackaging
ACR439341000
Catalog number ACR439341000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7250-53-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown to pale gray
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.0 to ≤103.0%
Assay (HPLC)≥97.0%
Melting Point (clear melt)295.0-305.0?C
Chiral derivatization reagent
RUO – Research Use Only