Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR43678050050 gPlastic bottle
Catalog number ACR436780500
Price (MYR)
1,457.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,457.00
Add to cart
Specifications
Boiling Point2730.0°C
Chemical Name or MaterialNickel
Melting Point1455.0°C
CAS7440-02-0
Health Hazard 1GHS Signal Word: Danger
View more
For ultra trace analysis
RUO – Research Use Only