4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%
4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%
4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%
4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%
Thermo Scientific Chemicals
4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%
CAS: 931-40-8 | C4H6O4 | 118.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR43617025025 gGlass bottle
Catalog number ACR436170250
Price (MYR)
1,676.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,676.00
Add to cart
Specifications
Chemical Name or Material4-(Hydroxymethyl)-1, 3-dioxolan-2-one
CAS931-40-8
CAS Min %88.0
CAS Max %100.0
Health Hazard 2GHS H Statement: