Check Order Status
3-Amino-6-(trifluoromethyl)-1H-indazole, 95%, Thermo Scientific™
Element Search
Structure Search
3-Amino-6-(trifluoromethyl)-1H-indazole, 95%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
Packaging:
Glass bottle
Catalog number
ACR435810050
Price (MYR)
346.86
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
346.86
Add to cart
Specifications
Chemical Name or Material
3-Amino-6-(trifluoromethyl)-1H-indazole
CAS
2250-55-7
CAS Min %
94.0
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0