3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific Chemicals
CAS: 61272-71-7 | C7H6BrN3 | 212.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR435760010
Price (MYR)
846.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
846.00
Add to cart
Chemical Identifiers
CAS1317-33-5
IUPAC Namedisulfanylidenemolybdenum
Molecular FormulaMoS2
InChI KeyCWQXQMHSOZUFJS-UHFFFAOYSA-N
SMILESS=[Mo]=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Grey to black
Assay>=98.0 % (MoS2)