3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR435740050
Price (MYR)
592.06
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
592.06
Add to cart
Chemical Identifiers
CAS5292-53-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.5340-1.5380 @ 20°C