3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR435740010
Price (MYR)
235.81
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
235.81
Add to cart
Specifications
Chemical Name or Material3-Amino-1H-indazole
CAS874-05-5
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0