4-(Trifluoromethyl)phenylacetylene, 97%
4-(Trifluoromethyl)phenylacetylene, 97%
4-(Trifluoromethyl)phenylacetylene, 97%
4-(Trifluoromethyl)phenylacetylene, 97%
Thermo Scientific Chemicals
4-(Trifluoromethyl)phenylacetylene, 97%
CAS: 705-31-7 | C9H5F3 | 170.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4353700505 gGlass bottle
Catalog number ACR435370050
Price (MYR)
2,012.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,012.00
Add to cart
Chemical Identifiers
CAS6000-43-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofA≥97.5% (NaOH titration)
Melting Point178-182?C
FormCrystalline powder