N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
Thermo Scientific Chemicals
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
CAS: 110-18-9 | C6H16N2 | 116.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR433831000100 mLAcroSeal™ Glass Bottle
Catalog number ACR433831000
Price (MYR)
733.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
733.00
Add to cart
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray to black
FormPowder
Total Metal Impurities0.1% max.