1,2-Dichloroethane, 99.8%, for HPLC, Thermo Scientific Chemicals
1,2-Dichloroethane, 99.8%, for HPLC, Thermo Scientific Chemicals
1,2-Dichloroethane, 99.8%, for HPLC, Thermo Scientific Chemicals
1,2-Dichloroethane, 99.8%, for HPLC, Thermo Scientific Chemicals
1,2-Dichloroethane, 99.8%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2-Dichloroethane, 99.8%, for HPLC, Thermo Scientific Chemicals
CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR433580010
Price (MYR)
922.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
922.00
EA
Add to cart
Chemical Identifiers
CAS108-91-8
IUPAC Namecyclohexanamine
Molecular FormulaC6H13N
InChI KeyPAFZNILMFXTMIY-UHFFFAOYSA-N
SMILESNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4580 to 1.4600 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
For HPLC
RUO – Research Use Only