2-Mercaptoimidazole, 98%, Thermo Scientific Chemicals
2-Mercaptoimidazole, 98%, Thermo Scientific Chemicals
2-Mercaptoimidazole, 98%, Thermo Scientific Chemicals
2-Mercaptoimidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Mercaptoimidazole, 98%, Thermo Scientific Chemicals
CAS: 872-35-5 | C3H4N2S | 100.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR433360010
Price (MYR)
590.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
590.00
Add to cart
Specifications
Chemical Name or Material2-Mercaptoimidazole
CAS872-35-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0