4-Ethynylaniline, 97%, Thermo Scientific Chemicals
4-Ethynylaniline, 97%, Thermo Scientific Chemicals
4-Ethynylaniline, 97%, Thermo Scientific Chemicals
4-Ethynylaniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Ethynylaniline, 97%, Thermo Scientific Chemicals
CAS: 14235-81-5 | C8H7N | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR432950050
Price (MYR)
1,895.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,895.00
Add to cart
Specifications
Chemical Name or Material4-Ethynylaniline
CAS14235-81-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0