Trifluoroacetic acid, 99+%, for HPLC, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, for HPLC, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, for HPLC, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, for HPLC, Thermo Scientific Chemicals
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
View all
Glass bottle
Catalog number ACR432291000
Price (MYR)
2,456.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,456.00
Add to cart
Chemical Identifiers
CAS2752-17-2
IUPAC Name2-(2-aminoethoxy)ethan-1-amine
Molecular FormulaC4H12N2O
InChI KeyGXVUZYLYWKWJIM-UHFFFAOYSA-N
SMILESNCCOCCN
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Refractive index1.4550 to 1.4580 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
For HPLC
RUO – Research Use Only