kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
CAS: 11114-20-8 | C24H36O25S2-2 | 788.647 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR431591000
Price (MYR)
3,401.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,401.00
Add to cart
Chemical Identifiers
CAS1306-19-0
IUPAC Namecadmium(2+) oxidandiide
Molecular FormulaCdO
InChI KeyCFEAAQFZALKQPA-UHFFFAOYSA-N
SMILES[O--].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.