kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
CAS: 11114-20-8 | C24H36O25S2-2 | 788.647 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR431590250
Price (MYR)
1,601.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,601.00
Add to cart
Chemical Identifiers
CAS8001-54-5
Molecular FormulaC9H13ClN
SMILES[Cl-].C[N+](C)(*)CC1=CC=CC=C1
Molecular Weight (g/mol)170.66
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid or thick gel
Infrared spectrumConforms
Water=<10.0 % (K.F.)
Total amines=<3.6 %
Appearance (Color)Colorless to white to yellow