Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
CAS: 106-92-3 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR430951000
Price (MYR)
415.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
415.00
Add to cart
Specifications
Chemical Name or MaterialAllyl glycidyl ether
CAS106-92-3
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0