p-Toluenesulfonyl isocyanate, 96%, AcroSeal™
p-Toluenesulfonyl isocyanate, 96%, AcroSeal™
p-Toluenesulfonyl isocyanate, 96%, AcroSeal™
p-Toluenesulfonyl isocyanate, 96%, AcroSeal™
Thermo Scientific Chemicals
p-Toluenesulfonyl isocyanate, 96%, AcroSeal™
CAS: 4083-64-1 | C8H7NO3S | 197.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR430761000100 mLAcroSeal™ Glass bottle
Catalog number ACR430761000
Price (MYR)
427.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
427.00
Add to cart
Chemical Identifiers
CAS10043-52-4
IUPAC Namecalcium dichloride
Molecular FormulaCaCl2
InChI KeyUXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<0.2 %
Barium (Ba)=<0.4 %
Lead (Pb)=<5 ppm
Mercury (Hg)=<1 ppm
Appearance (Form)Granules