Epichlorohydrin, 99%, AcroSeal™, Thermo Scientific Chemicals
Epichlorohydrin, 99%, AcroSeal™, Thermo Scientific Chemicals
Epichlorohydrin, 99%, AcroSeal™, Thermo Scientific Chemicals
Epichlorohydrin, 99%, AcroSeal™, Thermo Scientific Chemicals
Epichlorohydrin, 99%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Epichlorohydrin, 99%, AcroSeal™, Thermo Scientific Chemicals
CAS: 106-89-8 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR430711000
Price (MYR)
431.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
431.00
Add to cart
Chemical Identifiers
CAS3182-93-2
IUPAC Namehydrogen ethyl (2S)-2-amino-3-phenylpropanoate chloride
Molecular FormulaC11H16ClNO2
InChI KeyFPFQPLFYTKMCHN-PPHPATTJSA-N
SMILES[H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or needles
Appearance (Color)White
Melting point154°C to 157°C
Titration with HClO4>=98.5 %