2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
CAS: 40283-46-3 | C4H4N2O2S | 144.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR429570010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5153-40-2
IUPAC Name1-bromo-2,3,4,5,6-pentamethylbenzene
Molecular FormulaC11H15Br
InChI KeyXPDQRULPGCFCLX-UHFFFAOYSA-N
SMILESCC1=C(C)C(C)=C(Br)C(C)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)158.5-164.5°C
Appearance (Color)White
Assay (GC)≥97.5%