2-Aminothiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
2-Aminothiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
2-Aminothiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
2-Aminothiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminothiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
CAS: 40283-41-8 | C4H4N2O2S | 144.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR429480050
Price (MYR)
1,655.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,655.00
Add to cart
Specifications
Chemical Name or Material2-Aminothiazole-4-carboxylic acid
CAS40283-41-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0