Ethyl formate, 98+%, Extra Dry, AcroSeal™
Ethyl formate, 98+%, Extra Dry, AcroSeal™
Ethyl formate, 98+%, Extra Dry, AcroSeal™
Ethyl formate, 98+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Ethyl formate, 98+%, Extra Dry, AcroSeal™
CAS: 109-94-4 | C3H6O2 | 74.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR429241000100 mLAcroSeal™ Glass bottle
Catalog number ACR429241000
Price (MYR)
559.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
559.00
Add to cart
Specifications
Chemical Name or MaterialEthyl formate
CAS109-94-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapour.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a POISON CENTER or doctor/physician if you