Titanium(III) chloride, 20%w/v solution in 2N Hydrochloric acid,AcroSeal™
Titanium(III) chloride, 20%w/v solution in 2N Hydrochloric acid,AcroSeal™
Titanium(III) chloride, 20%w/v solution in 2N Hydrochloric acid,AcroSeal™
Titanium(III) chloride, 20%w/v solution in 2N Hydrochloric acid,AcroSeal™
Thermo Scientific Chemicals
Titanium(III) chloride, 20%w/v solution in 2N Hydrochloric acid,AcroSeal™
CAS: 7705-07-9 | Cl3Ti | 154.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR429151000100 mLAcroSeal™ Glass Bottle
Catalog number ACR429151000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS520-45-6
IUPAC Name3-acetyl-6-methyl-3,4-dihydro-2H-pyran-2,4-dione
Molecular FormulaC8H8O4
InChI KeyPGRHXDWITVMQBC-UHFFFAOYNA-N
SMILESCC(=O)C1C(=O)OC(C)=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 % (700°C, 2 h)
Appearance (Color)Cream to light yellow
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Melting point109°C to 112°C