Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
CAS: 7789-33-5 | BrI | 206.81 g/mol
Have Questions?
Catalog NumberdimQuantityPackaging
ACR428701000
Catalog number ACR428701000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS5122-95-2
IUPAC Name{[1,1'-biphenyl]-3-yl}boronic acid
Molecular FormulaC12H11BO2
InChI KeyGOXICVKOZJFRMB-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC(=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or granules
Assay (Aqueous acid-base Titration)≥97.5%
Assay (HPLC)≥97.5%
Proton NMRConforms to structure