Hydrogen chloride, 5 to 6N solution in 2-propanol, AcroSeal™, Thermo Scientific Chemicals
Hydrogen chloride, 5 to 6N solution in 2-propanol, AcroSeal™, Thermo Scientific Chemicals
Hydrogen chloride, 5 to 6N solution in 2-propanol, AcroSeal™, Thermo Scientific Chemicals
Hydrogen chloride, 5 to 6N solution in 2-propanol, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrogen chloride, 5 to 6N solution in 2-propanol, AcroSeal™, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR428608000
Price (MYR)
743.00
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
743.00
Add to cart
Chemical Identifiers
CAS79-06-1
IUPAC Nameprop-2-enamide
Molecular FormulaC3H5NO
InChI KeyHRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILESNC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Color of solution(10 % in water) Clear and colorless
Appearance (Color)White
Melting point82°C to 86°C
Water=<2.0 %