Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
Thermo Scientific Chemicals
Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™
CAS: 7550-45-0 | Cl4Ti | 189.71 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR428351000100 mLAcroSeal™ Glass Bottle
Catalog number ACR428351000
Price (MYR)
635.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
635.00
Add to cart
Chemical Identifiers
CAS7085-85-0
SpecificationsSpecification SheetSpecification Sheet
NMRConsistent with structure
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Lewis acid
RUO – Research Use Only