n-Hexadecyl palmitate, 95%, Thermo Scientific Chemicals
n-Hexadecyl palmitate, 95%, Thermo Scientific Chemicals
n-Hexadecyl palmitate, 95%, Thermo Scientific Chemicals
n-Hexadecyl palmitate, 95%, Thermo Scientific Chemicals
n-Hexadecyl palmitate, 95%, Thermo Scientific Chemicals
CAS: 540-10-3 | C32H64O2 | 480.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ACR428222500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7783-28-0
IUPAC Namephosphoric acid diamine
Molecular FormulaH9N2O4P
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILESN.N.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofA≥97.5% (US-sourced material)
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (UK-sourced material)
Identification (FTIR)Conforms (UK-sourced material)
FormCrystals or powder or crystalline powder or fused solid