L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
CAS: 6100-19-2 | C8H14K4O13 | 474.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ACR427995000
Price (MYR)
1,012.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,012.00
Add to cart
Chemical Identifiers
CAS97-85-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.3975-1.4015 @ 20?C