L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
CAS: 6100-19-2 | C8H14K4O13 | 474.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR427991000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-(+)-Tartaric acid dipotassium salt
CAS6100-19-2
Assay Percent Range97%
Molecular FormulaC8H14K4O13
MDL NumberMFCD00013065