Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
CAS: 23519-77-9 | C12H28O4Zr | 327.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR427931000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS60134-26-1
IUPAC Name(2R,3S)-2-(hydroxymethyl)-5-methoxyoxolan-3-ol
Molecular FormulaC6H12O4
InChI KeyNVGJZDFWPSOTHM-XCSHRFQENA-N
SMILESCOC1C[C@H](O)[C@@H](CO)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormViscous liquid
Assay (GC)≥88.0% (sum of isomers)
Optical Rotation22.0° to 40.0° (C=1 in water)
Refractive Index1.4665-1.4725 @ 20°C