7-Hydroxycoumarinyl-4-acetic acid, 97%, Thermo Scientific Chemicals
7-Hydroxycoumarinyl-4-acetic acid, 97%, Thermo Scientific Chemicals
7-Hydroxycoumarinyl-4-acetic acid, 97%, Thermo Scientific Chemicals
7-Hydroxycoumarinyl-4-acetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
7-Hydroxycoumarinyl-4-acetic acid, 97%, Thermo Scientific Chemicals
CAS: 6950-82-9 | C11H8O5 | 220.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR427900010
Price (MYR)
612.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
612.00
Add to cart
Chemical Identifiers
CAS1313-96-8
IUPAC Name[(dioxoniobio)oxy]dioxoniobium
Molecular FormulaNb2O5
InChI KeyZKATWMILCYLAPD-UHFFFAOYSA-N
SMILESO=[Nb](=O)O[Nb](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.