Phenethylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Phenethylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Phenethylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Phenethylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenethylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number ACR427801000
Price (MYR)
743.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
743.00
Add to cart
Specifications
Chemical Name or MaterialPhenethylmagnesium chloride
Name Note1M solution in THF
CAS90878-19-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/